siRNA Details
| Gene Symbol: Crygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109875.1 | NP_001103345.1 | 3 | 323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crygb| Gene Symbol: Crygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109875.1 | NP_001103345.1 | 3 | 323 | ||