siRNA Details
| Gene Symbol: Prr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104527.1 | NP_001097997.1 | 2 | 602 | ||
| GenBank mRNA | BC127529.1 | - | 2 | 602 | 7126230 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr15| Gene Symbol: Prr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104527.1 | NP_001097997.1 | 2 | 602 | ||
| GenBank mRNA | BC127529.1 | - | 2 | 602 | 7126230 | |