siRNA Details
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128194.2 | NP_001121666.1 | 4 | 971 | ||
GenBank mRNA | BC162055.1 | - | 4 | 971 | 7114289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128194.2 | NP_001121666.1 | 4 | 971 | ||
GenBank mRNA | BC162055.1 | - | 4 | 971 | 7114289 |