siRNA Details
| Gene Symbol: Fabp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017098.1 | NP_058794.1 | 2 | 236 | ||
| XM_008767638.1 | XP_008765860.1 | 1 | 308 | |||
| GenBank mRNA | L22788.1 | - | 2 | 236 | ||
| S52878.1 | - | 2 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp6| Gene Symbol: Fabp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017098.1 | NP_058794.1 | 2 | 236 | ||
| XM_008767638.1 | XP_008765860.1 | 1 | 308 | |||
| GenBank mRNA | L22788.1 | - | 2 | 236 | ||
| S52878.1 | - | 2 | 215 | |||