siRNA Details
| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100762.1 | NP_001094232.1 | 2 | 369 | ||
| XM_006246171.2 | XP_006246233.1 | 2 | 365 | |||
| GenBank mRNA | AJ549521.1 | - | 1 | 237 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Havcr2| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100762.1 | NP_001094232.1 | 2 | 369 | ||
| XM_006246171.2 | XP_006246233.1 | 2 | 365 | |||
| GenBank mRNA | AJ549521.1 | - | 1 | 237 | ||