siRNA Details
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115022.1 | NP_001108494.1 | 2 | 551 | ||
GenBank mRNA | BC158752.1 | - | 2 | 538 | 9102854 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115022.1 | NP_001108494.1 | 2 | 551 | ||
GenBank mRNA | BC158752.1 | - | 2 | 538 | 9102854 |