siRNA Details
| Gene Symbol: Wdr35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099340.1 | NP_001092810.1 | 15 | 1675 | ||
| XM_006239885.2 | XP_006239947.1 | 16 | 1724 | |||
| GenBank mRNA | EF613262.1 | - | 15 | 1675 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr35l| Gene Symbol: Wdr35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099340.1 | NP_001092810.1 | 15 | 1675 | ||
| XM_006239885.2 | XP_006239947.1 | 16 | 1724 | |||
| GenBank mRNA | EF613262.1 | - | 15 | 1675 | ||