siRNA Details
| Gene Symbol: Dmxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751784.3 | XP_003751832.1 | 2 | 491 | ||
| XM_006254710.2 | XP_006254772.1 | 2 | 491 | |||
| XM_006254711.2 | XP_006254773.1 | 2 | 491 | |||
| XM_006254712.2 | XP_006254774.1 | 2 | 491 | |||

Dmxl1
Genomic Map