siRNA Details
| Gene Symbol: Fam219a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109616.1 | NP_001103086.1 | 4, 5 | 565 | ||
| XM_006238100.2 | XP_006238162.1 | 4, 5 | 775 | |||
| XM_006238101.2 | XP_006238163.1 | 4, 5 | 776 | |||
| XM_006238102.2 | XP_006238164.1 | 4, 5 | 722 | |||
| XM_006238103.2 | XP_006238165.1 | 4, 5 | 724 | |||

Fam219a
Genomic Map