siRNA Details
| Gene Symbol: Sin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115038.1 | NP_001108510.1 | 6 | 832 | ||
| GenBank mRNA | BC158844.1 | - | 6 | 745 | 7442664 | |
| FQ220173.1 | - | 6 | 755 | |||
| FQ220358.1 | - | 6 | 755 | |||
| FQ220509.1 | - | 6 | 755 | |||
| FQ220565.1 | - | 6 | 755 | |||
| FQ220717.1 | - | 6 | 755 | |||
| FQ229609.1 | - | 6 | 755 | |||
| FQ229881.1 | - | 6 | 755 | |||

Sin3b
Genomic Map