siRNA Details
| Gene Symbol: Ptcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109665.1 | NP_001103135.1 | 4 | 810 | ||
| XM_006248871.1 | XP_006248933.1 | 4 | 839 | |||
| XM_006248873.1 | XP_006248935.1 | 3 | 698 | |||
| GenBank mRNA | BC129116.1 | - | 4 | 748 | 7113830 | |

Ptcd1
Genomic Map