siRNA Details
Gene Symbol: Tmem158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057212.1 | NP_476560.1 | 1 | 361 | ||
GenBank mRNA | AB028891.1 | - | 1 | 361 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057212.1 | NP_476560.1 | 1 | 361 | ||
GenBank mRNA | AB028891.1 | - | 1 | 361 |