siRNA Details
| Gene Symbol: Piga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108816.2 | NP_001102286.1 | 5 | 716 | ||
| XM_006256870.2 | XP_006256932.1 | 6 | 2966 | |||
| XM_006256873.2 | XP_006256935.1 | 6 | 1544 | |||
| XM_006256875.2 | XP_006256937.1 | 6 | 3086 | |||
| XM_006256876.1 | XP_006256938.1 | 6 | 1600 | |||
| XM_008773175.1 | XP_008771397.1 | 5 | 4264 | |||
| XM_008773176.1 | XP_008771398.1 | 6 | 3947 | |||
| XM_008773177.1 | XP_008771399.1 | 6 | 3535 | |||
| XM_008773178.1 | XP_008771400.1 | 6 | 4003 | |||
| XM_008773179.1 | XP_008771401.1 | 6 | 3011 | |||
| XM_008773180.1 | XP_008771402.1 | 6 | 3067 | |||
| XM_008773181.1 | XP_008771403.1 | 6 | 2068 | |||

Piga
Genomic Map