siRNA Details
| Gene Symbol: Ndc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126270.1 | NP_001119742.1 | 7 | 815 | ||
| XM_006245657.2 | XP_006245719.1 | 16 | 1853 | |||
| XM_008767423.1 | XP_008765645.1 | 16 | 1865 | |||
| GenBank mRNA | BC161935.1 | - | 16 | 1800 | 9022458 | |

Ndc80
Genomic Map