siRNA Details
| Gene Symbol: Smim22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108266.1 | NP_001101736.1 | 4 | 462 | ||
| XM_006245806.2 | XP_006245868.1 | 4 | 1095 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim22| Gene Symbol: Smim22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108266.1 | NP_001101736.1 | 4 | 462 | ||
| XM_006245806.2 | XP_006245868.1 | 4 | 1095 | |||