siRNA Details
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031117.2 | NP_112379.1 | 9 | 1230 | ||
GenBank mRNA | BC087671.1 | - | 5 | 634 | 7316987 | |
M29293.1 | - | 9 | 1230 | |||
M29294.1 | - | 5 | 534 |
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270712.1 | NP_001257641.1 | 10 | 1254 | ||
NM_130738.2 | NP_570094.1 | 9 | 1119 |
