siRNA Details
Gene Symbol: RGD1305743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108060.2 | NP_001101530.1 | 1 | 1322 | ||
GenBank mRNA | BC168849.1 | - | 1 | 1322 | 7374840 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1305743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108060.2 | NP_001101530.1 | 1 | 1322 | ||
GenBank mRNA | BC168849.1 | - | 1 | 1322 | 7374840 |