siRNA Details
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106076.1 | NP_001099546.1 | 4 | 508 | ||
XM_006252876.2 | XP_006252938.1 | 4 | 522 | |||
GenBank mRNA | BC161928.1 | - | 4 | 522 | 8374450 | |
FQ220757.1 | - | 4 | 332 | |||
FQ232202.1 | - | 4 | 331 | |||
FQ233163.1 | - | 4 | 331 |
