siRNA Details
| Gene Symbol: Cope | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106076.1 | NP_001099546.1 | 2 | 352 | ||
| XM_006252876.2 | XP_006252938.1 | 2 | 366 | |||
| GenBank mRNA | BC161928.1 | - | 2 | 366 | 8374450 | |
| FQ220757.1 | - | 2 | 176 | |||
| FQ232202.1 | - | 2 | 175 | |||
| FQ233163.1 | - | 2 | 175 | |||

Cope
Genomic Map