siRNA Details
| Gene Symbol: Wdr92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127559.2 | NP_001121031.1 | 6 | 706 | ||
| GenBank mRNA | BC162042.1 | - | 6 | 706 | 7105425 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr92| Gene Symbol: Wdr92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127559.2 | NP_001121031.1 | 6 | 706 | ||
| GenBank mRNA | BC162042.1 | - | 6 | 706 | 7105425 | |