siRNA Details
| Gene Symbol: Ms4a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231100.2 | XP_006231162.1 | 7 | 1810 | ||
| XM_008760272.1 | XP_008758494.1 | 7 | 1703 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a12| Gene Symbol: Ms4a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231100.2 | XP_006231162.1 | 7 | 1810 | ||
| XM_008760272.1 | XP_008758494.1 | 7 | 1703 | |||