siRNA Details
| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108950.2 | NP_001102420.2 | 3 | 228 | ||
| XM_006233171.1 | XP_006233233.1 | 3 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnat2| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108950.2 | NP_001102420.2 | 3 | 228 | ||
| XM_006233171.1 | XP_006233233.1 | 3 | 261 | |||