siRNA Details
| Gene Symbol: Mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115025.1 | NP_001108497.1 | 3 | 773 | ||
| GenBank mRNA | BC158877.1 | - | 3 | 773 | 7934371 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbd2| Gene Symbol: Mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115025.1 | NP_001108497.1 | 3 | 773 | ||
| GenBank mRNA | BC158877.1 | - | 3 | 773 | 7934371 | |