siRNA Details
| Gene Symbol: Hdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014151.2 | NP_001014173.2 | 2 | 162 | ||
| GenBank mRNA | AY539873.1 | - | 2 | 162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdhd2| Gene Symbol: Hdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014151.2 | NP_001014173.2 | 2 | 162 | ||
| GenBank mRNA | AY539873.1 | - | 2 | 162 | ||