siRNA Details
| Gene Symbol: Tmem125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107967.2 | NP_001101437.2 | 1 | 853 | ||
| GenBank mRNA | BC086537.1 | - | 1 | 853 | 7106780 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem125| Gene Symbol: Tmem125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107967.2 | NP_001101437.2 | 1 | 853 | ||
| GenBank mRNA | BC086537.1 | - | 1 | 853 | 7106780 | |