siRNA Details
Gene Symbol: Tmem125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107967.2 | NP_001101437.2 | 1 | 723 | ||
GenBank mRNA | BC086537.1 | - | 1 | 723 | 7106780 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107967.2 | NP_001101437.2 | 1 | 723 | ||
GenBank mRNA | BC086537.1 | - | 1 | 723 | 7106780 |