siRNA Details
| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134994.1 | NP_001128466.1 | 3 | 256 | ||
| GenBank mRNA | BC168695.1 | - | 3 | 145 | 7938123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpk1| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134994.1 | NP_001128466.1 | 3 | 256 | ||
| GenBank mRNA | BC168695.1 | - | 3 | 145 | 7938123 | |