siRNA Details
| Gene Symbol: Wbscr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109499.2 | NP_001102969.1 | 5 | 691 | ||
| XM_006249190.2 | XP_006249252.1 | 6 | 777 | |||
| XM_006249191.2 | XP_006249253.1 | 6 | 777 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr27| Gene Symbol: Wbscr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109499.2 | NP_001102969.1 | 5 | 691 | ||
| XM_006249190.2 | XP_006249252.1 | 6 | 777 | |||
| XM_006249191.2 | XP_006249253.1 | 6 | 777 | |||