siRNA Details
| Gene Symbol: LOC689065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109521.1 | NP_001102991.1 | 1, 2 | 339 | ||
| XM_006230905.2 | XP_006230967.1 | 2, 3 | 924 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689065| Gene Symbol: LOC689065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109521.1 | NP_001102991.1 | 1, 2 | 339 | ||
| XM_006230905.2 | XP_006230967.1 | 2, 3 | 924 | |||