siRNA Details
| Gene Symbol: Pink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106694.1 | NP_001100164.1 | 2 | 500 | ||
| GenBank mRNA | BC169047.1 | - | 2 | 494 | 7190543 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pink1| Gene Symbol: Pink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106694.1 | NP_001100164.1 | 2 | 500 | ||
| GenBank mRNA | BC169047.1 | - | 2 | 494 | 7190543 | |