siRNA Details
| Gene Symbol: Defa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033074.1 | NP_001028246.1 | 2 | 202 | ||
| XM_008771364.1 | XP_008769586.1 | 3 | 403 | |||
| XM_008771365.1 | XP_008769587.1 | 3 | 400 | |||
| GenBank mRNA | AY623754.1 | - | 2 | 202 | ||

Defa10
Genomic Map