siRNA Details
| Gene Symbol: Vps9d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107440.2 | NP_001100910.2 | 3 | 257 | ||
| XM_006255767.1 | XP_006255829.1 | 3 | 303 | |||
| XM_006255768.2 | XP_006255830.1 | 3 | 303 | |||
| XM_008772613.1 | XP_008770835.1 | 3 | 772 | |||
| GenBank mRNA | BC166767.1 | - | 3 | 257 | 7933213 | |
| FQ221344.1 | - | 3 | 268 | |||

Vps9d1
Genomic Map