siRNA Details
| Gene Symbol: Chtf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001194951.1 | NP_001181880.1 | 3 | 770 | ||
| GenBank mRNA | BC166818.1 | - | 1 | 423 | 8367310 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chtf8| Gene Symbol: Chtf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001194951.1 | NP_001181880.1 | 3 | 770 | ||
| GenBank mRNA | BC166818.1 | - | 1 | 423 | 8367310 | |