siRNA Details
| Gene Symbol: Tbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127201.1 | NP_001120673.1 | 3 | 432 | ||
| XM_006227721.2 | XP_006227783.1 | 3 | 465 | |||
| XM_006227722.2 | XP_006227784.1 | 3 | 389 | |||
| XM_008758627.1 | XP_008756849.1 | 4 | 432 | |||
| GenBank mRNA | CF109177.1 | - | 2 | 124 | ||
| FQ213058.1 | - | 2 | 84 | |||
| FQ220549.1 | - | 2 | 84 | |||
| FQ234587.1 | - | 3 | 450 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011603, AK078547, AK049975, AK005604, BC033926, and others.

Tbpl1
Genomic Map