siRNA Details
| Gene Symbol: Scaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037347.2 | NP_001032424.2 | 20 | 3544 | ||
| XM_006248024.2 | XP_006248086.1 | 20 | 3724 | |||
| XM_006248025.2 | XP_006248087.1 | 20 | 3658 | |||
| XM_008768558.1 | XP_008766780.1 | 20 | 3712 | |||
| GenBank mRNA | U49058.1 | - | 19 | 3419 | ||

Scaf4
Genomic Map