siRNA Details
| Gene Symbol: Sln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013247.1 | NP_001013265.1 | 2 | 180 | ||
| GenBank mRNA | AY456000.1 | - | 2 | 180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sln| Gene Symbol: Sln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013247.1 | NP_001013265.1 | 2 | 180 | ||
| GenBank mRNA | AY456000.1 | - | 2 | 180 | ||