siRNA Details
| Gene Symbol: LOC680039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109388.1 | NP_001102858.1 | 6 | 3864 | ||
| XM_006251140.2 | XP_006251202.1 | 6 | 3906 | |||
| XM_006251141.2 | XP_006251203.1 | 6 | 3906 | |||
| XM_008770360.1 | XP_008768582.1 | 4 | 3627 | |||

LOC680039
Genomic Map