siRNA Details
| Gene Symbol: Fosl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012954.1 | NP_037086.1 | 3 | 394 | ||
| GenBank mRNA | U18913.1 | - | 3 | 394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fosl2| Gene Symbol: Fosl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012954.1 | NP_037086.1 | 3 | 394 | ||
| GenBank mRNA | U18913.1 | - | 3 | 394 | ||