siRNA Details
| Gene Symbol: Chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109639.1 | NP_001103109.1 | 1 | 411 | ||
| GenBank mRNA | BC166542.1 | - | 1 | 356 | 7193817 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst14| Gene Symbol: Chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109639.1 | NP_001103109.1 | 1 | 411 | ||
| GenBank mRNA | BC166542.1 | - | 1 | 356 | 7193817 | |