siRNA Details
| Gene Symbol: Dab2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138710.3 | NP_619724.3 | 4, 5 | 679 | ||
| XM_006234057.2 | XP_006234119.1 | 3, 4 | 666 | |||
| XM_008761697.1 | XP_008759919.1 | 1, 2 | 1077 | |||
| XM_008761698.1 | XP_008759920.1 | 1, 2 | 1077 | |||
| XM_008761699.1 | XP_008759921.1 | 1, 2 | 1078 | |||

Dab2ip
Genomic Map