siRNA Details
| Gene Symbol: Minpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019263.1 | NP_062136.1 | 5 | 1394 | ||
| XM_008760326.1 | XP_008758548.1 | 5 | 971 | |||
| XM_008760327.1 | XP_008758549.1 | 3 | 1228 | |||
| GenBank mRNA | AF012714.2 | - | 5 | 1394 | ||
| CO388212.1 | - | 2 | 371 | |||

Minpp1
Genomic Map