siRNA Details
| Gene Symbol: Itih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107291.2 | NP_001100761.2 | 3 | 165 | ||
| XM_008770995.1 | XP_008769217.1 | 3 | 210 | |||
| GenBank mRNA | BC166831.1 | - | 3 | 165 | 8374222 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itih1| Gene Symbol: Itih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107291.2 | NP_001100761.2 | 3 | 165 | ||
| XM_008770995.1 | XP_008769217.1 | 3 | 210 | |||
| GenBank mRNA | BC166831.1 | - | 3 | 165 | 8374222 | |