siRNA Details
| Gene Symbol: Tprg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110318.2 | NP_001103788.1 | 4 | 806 | ||
| NM_001271085.1 | NP_001258014.1 | 3 | 652 | |||
| GenBank mRNA | BC105914.1 | - | 3 | 513 | ||
| BC160865.1 | - | 4 | 655 | 7324838 | ||
| EU220430.1 | - | 4 | 806 | |||

Tprg1l
Genomic Map