siRNA Details
| Gene Symbol: Hist3h2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111127.1 | NP_001104597.1 | 1 | 579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist3h2ba| Gene Symbol: Hist3h2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111127.1 | NP_001104597.1 | 1 | 579 | ||