siRNA Details
| Gene Symbol: Slc41a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108855.1 | NP_001102325.1 | 6 | 1768 | ||
| XM_008769486.1 | XP_008767708.1 | 5 | 891 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc41a1| Gene Symbol: Slc41a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108855.1 | NP_001102325.1 | 6 | 1768 | ||
| XM_008769486.1 | XP_008767708.1 | 5 | 891 | |||