siRNA Details
| Gene Symbol: Sct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022670.2 | NP_073161.1 | 3 | 205 | ||
| XM_006230518.2 | XP_006230580.1 | 4 | 453 | |||
| GenBank mRNA | M31495.1 | - | 3 | 228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sct| Gene Symbol: Sct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022670.2 | NP_073161.1 | 3 | 205 | ||
| XM_006230518.2 | XP_006230580.1 | 4 | 453 | |||
| GenBank mRNA | M31495.1 | - | 3 | 228 | ||