siRNA Details
| Gene Symbol: Car14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109655.2 | NP_001103125.1 | 4, 5 | 406 | ||
| XM_006232988.2 | XP_006233050.1 | 5, 6 | 951 | |||
| XM_006232989.2 | XP_006233051.1 | 5, 6 | 929 | |||
| XM_006232990.2 | XP_006233052.1 | 5, 6 | 911 | |||
| GenBank mRNA | EF187253.1 | - | 4, 5 | 406 | ||

Car14
Genomic Map