siRNA Details
| Gene Symbol: Mif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031051.1 | NP_112313.1 | 3 | 385 | ||
| GenBank mRNA | BC061545.1 | - | 3 | 369 | 6920605 | |
| FQ210474.1 | - | 3 | 367 | |||
| FQ213295.1 | - | 3 | 354 | |||
| FQ217414.1 | - | 3 | 365 | |||
| FQ218686.1 | - | 3 | 354 | |||
| FQ222835.1 | - | 3 | 381 | |||
| FQ223656.1 | - | 3 | 342 | |||
| FQ229019.1 | - | 3 | 365 | |||
| FQ229084.1 | - | 3 | 365 | |||
| S73424.1 | - | 3 | 350 | |||
| U20999.1 | - | 3 | 356 | |||
| U62326.1 | - | 3 | 385 | |||
| Gene Symbol: LOC103694877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773846.1 | XP_008772068.1 | 3 | 549 | ||

Mif
Genomic Map