siRNA Details
| Gene Symbol: Hsp90ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004082.3 | NP_001004082.3 | 7 | 1259 | ||
| GenBank mRNA | AY695392.1 | - | 7 | 1259 | ||
| AY695393.1 | - | 7 | 1247 | |||
| AY724481.1 | - | 7 | 1230 | |||
| BC082009.1 | - | 7 | 1285 | 7131767 | ||
| FQ223952.1 | - | 3 | 385 | |||
| FQ231832.1 | - | 1 | 67 | |||
| S45392.1 | - | 7 | 1291 | |||

Hsp90ab1
Genomic Map