siRNA Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126296.2 | NP_001119768.2 | 4 | 606 | ||
| XM_006236464.2 | XP_006236526.1 | 4 | 625 | |||
| XM_006236465.2 | XP_006236527.1 | 4 | 625 | |||
| XM_006236466.2 | XP_006236528.1 | 4 | 625 | |||
| GenBank mRNA | BC161978.1 | - | 4 | 606 | 9029930 | |

Tra2a
Genomic Map